3,4,5-Trimethoxyphenol 98.5+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Antiarol

18914-0050 18914-0250 18914-1000
TS18914-0050EA 89 USD
TS18914-0050 TS18914-0250 TS18914-1000
3,4,5-Trimethoxyphenol 98.5+%
3,4,5-Trimethoxyphenol
Formula: C₉H₁₂O₄
MW: 184.19
Boiling Pt: 283
Melting Pt: 146…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00008389
CAS Number: 642-71-7
EINECS: 211-387-2

Order Now

Specification Test Results

Appearance White to yellow to beige to brown Powder or crystals or needles
Infrared spectrum Conforms
Melting point 143 to 149 °C
GC ≥98.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR