2,3,4,5,6-Pentafluorostyrene 97% stabilized

Supplier: Thermo Scientific Chemicals
Warning

18847-0050 18847-0250
TS18847-0050EA 61 USD
TS18847-0050 TS18847-0250
2,3,4,5,6-Pentafluorostyrene 97% stabilized
2,3,4,5,6-Pentafluorostyrene
Formula: C₈H₃F₅
Boiling Pt: 62…63 °C (50 mmHg)
Density: 1.406 g/cm³
Flash Pt: 34 °C
Storage Temperature: Freezer
MDL Number: MFCD00000300
CAS Number: 653-34-9
EINECS: 211-500-5
UN: 1993

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4450 to 1.4470 (20°C, 589 nm)
GC ≥96.0 %
Stabilizer 0.01 to 0.1 % p-tert-Butylcatechol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR