Isoindolin-1-one

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,3-Dihydro-1H-isoindol-1-one, 1-Isoindolinone

46518-0050 465180010
TS46518-0050EA 186 USD
TS46518-0050 TS465180010
Isoindolin-1-one
Isoindolin-1-one
Formula: C₈H₇NO
MW: 133.15 g/mol
Melting Pt: 150…160 °C
Storage Temperature: Ambient
MDL Number: MFCD03085939
CAS Number: 480-91-1

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 149 to 153 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR