7-Hydroxyindole 95%

Supplier: Thermo Scientific Chemicals
Danger

46229-2500 462290010
TS46229-2500EA 142 USD
TS46229-2500 TS462290010
7-Hydroxyindole 95%
7-Hydroxyindole
Formula: C₈H₇NO
Melting Pt: 85…87 °C
Storage Temperature: Freezer
MDL Number: MFCD00152102
CAS Number: 2380-84-9

Order Now

Specification Test Results

Appearance Yellow to brown Solid or crystalline solid
Infrared spectrum Conforms
Melting point 83 to 88 °C
GC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR