Raltegravir potassium salt

Supplier: Thermo Scientific Chemicals
Danger

46216-0010 46216-2500
TS46216-0010EA 631 USD
TS46216-0010 TS46216-2500
Raltegravir potassium salt
Raltegravir potassium salt
Formula: C₂₀H₂₀FKN₆O₅
Melting Pt: 155…157 °C
Storage Temperature: Freezer
MDL Number: MFCD12031642
CAS Number: 871038-72-1

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR