1,8-Diazafluoren-9-one

Supplier: Thermo Scientific Chemicals
Warning

46131-0010 46131-0050
TS46131-0010EA 380 USD
TS46131-0010 TS46131-0050
1,8-Diazafluoren-9-one
1,8-Diazafluoren-9-one
Formula: C₁₁H₆N₂O
Melting Pt: 229…233 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00082335
CAS Number: 54078-29-4

Order Now

Specification Test Results

Appearance Yellow to orange Powder
Infrared spectrum Conforms
Loss on drying ≤2 %
Dye content ≥70.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR