2,3,5,6-Tetrafluorophenol 98%

Supplier: Thermo Scientific Chemicals
Danger

18839-0050 18839-0250
TS18839-0050EA 60 USD
TS18839-0050 TS18839-0250
2,3,5,6-Tetrafluorophenol 98%
2,3,5,6-Tetrafluorophenol
Formula: C₆H₂F₄O
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: 33…38 °C
Flash Pt: 79 °C
MDL Number: MFCD00002157
CAS Number: 769-39-1
EINECS: 212-209-6
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Appearance White Low melting crystalline solid
Infrared spectrum Conforms
Melting point 33 to 38 °C
GC ≥97.5 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR