Biacore maintenance kit, Cytiva
Supplier: Cytiva
29394519
29394521
103871-802EA
526.15
USD
103871-802
103871-804
103871-806
Biacore maintenance kit, Cytiva
Protein Purification
Protein Purification Equipment
Biacore maintenance kits are convenient kits for proper instrument maintenance.
Shelf life: Sufficient for 6 months normal usage except for type 2 which wil be sufficient for 3 to 4 months usage.
Información para pedidos: The use of the CAPture kits in Biacore systems requires an Amine Coupling Kit with Sensor Chip CM5 CM4 CM3 or C. Not for use with Biacore Flexchip unless otherwise noted.