Levetiracetam

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-2-(2-Oxopyrrolidin-1-yl)butanamide, (S)-2-(2-Oxo-1-pyrrolidinyl)butyramide

46212-0050 46212-0250
TS46212-0050EA 84 USD
TS46212-0050 TS46212-0250
Levetiracetam
Levetiracetam
Formula: C₈H₁₄N₂O₂
MW: 170.21 g/mol
Melting Pt: 117 °C
Storage Temperature: Freezer
MDL Number: MFCD03265610
CAS Number: 102767-28-2

Order Now

Specification Test Results

Appearance White to almost white Powder
Infrared spectrum Conforms
Melting point 116 to 120 °C
Loss on drying ≤0.5 %
HPLC ≥97.5 %
Specific optical rotation -76° to -82° (20 °C, 589 nm) (2 % in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR