3-Amino-1-methyl-1H-pyrazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
45673-0050
45673-0250
TS45673-0050EA
49
USD
TS45673-0050
TS45673-0250
3-Amino-1-methyl-1H-pyrazole 97%
3-Amino-1-methyl-1H-pyrazole
Formula:
C₄H₇N₃ Boiling Pt: 93…94 °C (.5 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00466340 CAS Number: 1904-31-0 |
Specification Test Results
Appearance | Clear colorless to yellow to green to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5400 to 1.5450 (20°C, 589 nm) |
Water | ≤1.5 % |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...