5-Quinolinecarboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
45476-0010
45476-0050
TS45476-0010EA
139
USD
TS45476-0010
TS45476-0050
5-Quinolinecarboxylic acid 97%
5-Quinolinecarboxylic acid
Formula:
C₁₀H₇NO₂ MW: 173.17 g/mol Melting Pt: 337…341 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047608 CAS Number: 7250-53-5 |
Specification Test Results
Appearance | White to off-white to light brown Solid |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...