5-Aminopicolinic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Aminopyridine-2-carboxylic acid, 5-Amino-2-pyridinecarboxylic acid
45098-0010
45098-0050
TS45098-0010EA
115
USD
TS45098-0010
TS45098-0050
5-Aminopicolinic acid 98%
5-Aminopicolinic acid
Formula:
C₆H₆N₂O₂ MW: 138.13 g/mol Melting Pt: 215…217 °C Storage Temperature: Ambient |
MDL Number:
MFCD02684600 CAS Number: 24242-20-4 |
Specification Test Results
Appearance | White to off-white to light yellow Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...