2-Aminopyridine-6-methanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (6-Aminopyridin-2-yl)methanol

45004-0050 450040010
TS45004-0050EA 275 USD
TS45004-0050 TS450040010
2-Aminopyridine-6-methanol 97%
2-Aminopyridine-6-methanol
Formula: C₆H₈N₂O
MW: 124.14 g/mol
Melting Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD09863097
CAS Number: 79651-64-2

Order Now

Specification Test Results

Appearance Off-white to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 89 to 95 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR