2-Chloro-4-ethynylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

44998-0010 44998-0050
TS44998-0010EA 219 USD
TS44998-0010 TS44998-0050
2-Chloro-4-ethynylpyridine 97%
2-Chloro-4-ethynylpyridine
Formula: C₇H₄ClN
Boiling Pt: 373 °C (760 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD13189742
CAS Number: 945717-09-9

Order Now

Specification Test Results

Appearance White to light brown to green Crystalline powder or crystals
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR