5-Amino-2-(methylthio)pyrimidine 96%

Supplier: Thermo Scientific Chemicals
Warning

44982-2500 449820010
TS44982-2500EA 119 USD
TS44982-2500 TS449820010
5-Amino-2-(methylthio)pyrimidine 96%
5-Amino-2-(methylthio)pyrimidine
Formula: C₅H₇N₃S
Storage Temperature: Ambient
MDL Number: MFCD10697095
CAS Number: 42382-46-7

Order Now

Specification Test Results

Appearance Orange-red to red Powder
Infrared spectrum Conforms
HPLC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR