3-Amino-5-chloropyridine 95%

Supplier: Thermo Scientific Chemicals
Danger

44890-0010 44890-0050
TS44890-0010EA 127 USD
TS44890-0010 TS44890-0050
3-Amino-5-chloropyridine 95%
3-Amino-5-chloropyridine
Formula: C₅H₅ClN₂
Melting Pt: 51…56 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03701386
CAS Number: 22353-34-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Cream to beige to light brown Powder
Infrared spectrum Conforms
Melting point 71 to 75 °C
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR