3-Amino-4-iodopyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Iodo-3-pyridylamine, 4-Iodopyridin-3-amine, 4-Iodo-pyridin-3-ylamine

44884-0010 44884-0050
TS44884-0010EA 141 USD
TS44884-0010 TS44884-0050
3-Amino-4-iodopyridine 98%
3-Amino-4-iodopyridine
Formula: C₅H₅IN₂
MW: 220.01
Melting Pt: 65…70 °C
Storage Temperature: Ambient
MDL Number: MFCD04971334
CAS Number: 105752-11-2

Order Now

Specification Test Results

Appearance Off-white to beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 60 to 70 °C
GC ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR