2-Fluoro-4-pyridineboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Fluoropyridin-4-yl)boronic acid, 2-Fluoropyridine-4-boronic acid, 2-Fluoro-4-pyridinylboronic acid

44873-0010 44873-0050
TS44873-0010EA 144 USD
TS44873-0010 TS44873-0050
2-Fluoro-4-pyridineboronic acid 98%
2-Fluoro-4-pyridineboronic acid
Formula: C₅H₅BFNO₂
MW: 140.91 g/mol
Melting Pt: 212 °C
Storage Temperature: Ambient
MDL Number: MFCD04112534
CAS Number: 401815-98-3

Order Now

Specification Test Results

Appearance Light yellow to light brown Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR