Piperidine-2,4-dione 97%

Supplier: Thermo Scientific Chemicals
Warning

44673-0010 44673-0050
TS44673-0010EA 156 USD
TS44673-0010 TS44673-0050
Piperidine-2,4-dione 97%
Piperidine-2,4-dione
Formula: C₅H₇NO₂
Melting Pt: 99.1…100.1 °C
Storage Temperature: Ambient
MDL Number: MFCD08704814
CAS Number: 50607-30-2

Order Now

Specification Test Results

Appearance White to yellow to light brown Powder or solid
Infrared spectrum Conforms
Melting point 98 to 104 °C
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR