Methyl-2,4-dichloropyrimidine-6-carboxylate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,6-Dichloro-4-pyrimidinecarboxylic acid methyl ester, 2-(3,4-dichlorophenyl)-4-methyl-5-thiazolecarboxylic acid, Methyl 2,6-dichloro-4-pyrimidinecarboxylate, Methyl 2,6-dichloropyrimidine-4-carboxylate

44658-0050 44658-0250
TS44658-0050EA 158 USD
TS44658-0050 TS44658-0250
Methyl-2,4-dichloropyrimidine-6-carboxylate 98%
Methyl-2,4-dichloropyrimidine-6-carboxylate
Formula: C₆H₄Cl₂N₂O₂
MW: 207.02 g/mol
Melting Pt: 55…56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01941317
CAS Number: 6299-85-0

Order Now

Specification Test Results

Appearance White to yellow to light brown Powder
Infrared spectrum Conforms
Melting point 53 to 59 °C
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR