1-Boc-3-fluoro-4-piperidone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Piperidinecarboxylic acid, 3-fluoro-4-oxo-, 1,1-dimethylethyl ester, tert-Butyl 3-Fluoro-4-oxo-1-piperidinecarboxylate, tert-Butyl 3-fluoro-4-oxopiperidine-1-carboxylate, 3-Fluoro-4-oxopiperidine-1-carboxylic acid tert-butyl ester, 1-(tert-Butoxycarbonyl)-3-fluoro-4-piperidone, 3-Fluoro-4-oxo-1-piperidinecarboxylic acid tert-Butyl ester, 1-Boc-3-fluoro-4-piperidone

44651-0010 44651-0050
TS44651-0010EA 209 USD
TS44651-0010 TS44651-0050
1-Boc-3-fluoro-4-piperidone 97%
1-Boc-3-fluoro-4-piperidone
Formula: C₁₀H₁₆FNO₃
MW: 217.24 g/mol
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD10566554
CAS Number: 211108-50-8

Order Now

Specification Test Results

Appearance White to cream to light brown Powder or solid
Infrared spectrum Conforms
Melting point 73 to 78 °C
Water ≤2.5 %
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR