4-(Methylamino)phenylboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

44515-0010 44515-0050
TS44515-0010EA 256 USD
TS44515-0010 TS44515-0050
4-(Methylamino)phenylboronic acid pinacol ester 97%
4-(Methylamino)phenylboronic acid pinacol ester
Formula: C₁₃H₂₀BNO₂
MW: 233.12 g/mol
Melting Pt: 65…66 °C
Storage Temperature: Ambient
MDL Number: MFCD06795668
CAS Number: 845870-55-5

Order Now

Specification Test Results

Appearance White to brown to purple Solid
Infrared spectrum Conforms
Melting point 63 to 67 °C
GC ≥96 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR