4-Cyano-2-fluorophenylboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Cyano-2-fluorobenzeneboronic acid, pinacol ester, 3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 4-Cyano-2-fluorobenzeneboronic acid pinacol ester

44472-0010 44472-0050
TS44472-0010EA 302 USD
TS44472-0010 TS44472-0050
4-Cyano-2-fluorophenylboronic acid pinacol ester 97%
4-Cyano-2-fluorophenylboronic acid pinacol ester
Formula: C₁₃H₁₅BFNO₂
Melting Pt: 83…84 °C
Storage Temperature: Ambient
MDL Number: MFCD09998162
CAS Number: 1035235-29-0

Order Now

Specification Test Results

Appearance Colorless to white to yellow Solid
Infrared spectrum Conforms
Melting point 82 to 87 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR