6-(tert-Butoxycarbonylamino)pyridine-3-boronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Carbamic acid,N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridinyl]-,1,1-dimethylethyl ester, tert-Butyl (5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)carbamate, 2-(BOC-Amino)pyridine-5-boronic acid, pinacol ester

44466-0010 44466-0050
TS44466-0010EA 173 USD
TS44466-0010 TS44466-0050
6-(tert-Butoxycarbonylamino)pyridine-3-boronic acid pinacol ester 97%
6-(tert-Butoxycarbonylamino)pyridine-3-boronic acid pinacol ester
Formula: C₁₆H₂₅BN₂O₄
Melting Pt: 203…205 °C
Storage Temperature: Ambient
MDL Number: MFCD07781162
CAS Number: 910462-31-6

Order Now

Specification Test Results

Appearance White to off-white Solid
Infrared spectrum Conforms
Melting point 202 to 208 °C
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR