4-Thiazolecarboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Thiazole-4-carboxylic acid
44397-0010
44397-0050
TS44397-0010EA
70
USD
TS44397-0010
TS44397-0050
4-Thiazolecarboxylic acid 97%
4-Thiazolecarboxylic acid
Formula:
C₄H₃NO₂S MW: 129.1371 g/mol Melting Pt: 195…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00623589 CAS Number: 3973-08-8 |
Specification Test Results
Appearance | White to yellow to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 190 to 205 °C |
Loss on drying | ≤3 % |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...