2-Methylisonicotinic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methylpyridine-4-carboxylic acid

44349-0010 44349-0050
TS44349-0010EA 93 USD
TS44349-0010 TS44349-0050
2-Methylisonicotinic acid 97%
2-Methylisonicotinic acid
Formula: C₇H₇NO₂
MW: 137.14
Storage Temperature: Ambient
MDL Number: MFCD00513435
CAS Number: 4021-11-8

Order Now

Specification Test Results

Appearance White to brown Crystalline powder or solid
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR