4-Chloro-3-fluoropyridine 95%

Supplier: Thermo Scientific Chemicals
Warning

44316-0050 44316-0250
TS44316-0050EA 120 USD
TS44316-0050 TS44316-0250
4-Chloro-3-fluoropyridine 95%
4-Chloro-3-fluoropyridine
Formula: C₅H₃ClFN
MW: 131.54 g/mol
Boiling Pt: 142…143 °C (760 mmHg)
Melting Pt: 27…24 °C
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD03453233
CAS Number: 2546-56-7
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR