1-Butylimidazole 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Butyl-1H-imidazole, 1-Butyl-1H-imidazole, N-Butylimidazole

44277-0250 44277-1000
TS44277-0250EA 69 USD
TS44277-0250 TS44277-1000
1-Butylimidazole 98%
1-Butylimidazole
Formula: C₇H₁₂N₂
MW: 124.19 g/mol
Boiling Pt: 257 °C (760 mmHg)
Density: 0.945 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00042753
CAS Number: 4316-42-1
UN: 2690
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4790 to 1.4810 (20°C, 589 nm)
Water ≤0.5 %
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR