2-Chloro-4-fluoropyridine 97%

Supplier: Thermo Scientific Chemicals
Danger

44215-0010 44215-0050
TS44215-0010EA 110 USD
TS44215-0010 TS44215-0050
2-Chloro-4-fluoropyridine 97%
2-Chloro-4-fluoropyridine
Formula: C₅H₃ClFN
Boiling Pt: 140 °C (760 mmHg)
Density: 1.456 g/cm³
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD04039345
CAS Number: 34941-91-8
UN: 2929
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR