2-Amino-4-chloropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chloro-2-pyridylamine, 4-Chloropyridin-2-amine

44146-0050 44146-0250
TS44146-0050EA 67 USD
TS44146-0050 TS44146-0250
2-Amino-4-chloropyridine 97%
2-Amino-4-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Melting Pt: 130…132 °C
Storage Temperature: Ambient
MDL Number: MFCD04113820
CAS Number: 19798-80-2

Order Now

Specification Test Results

Appearance White to light brown Solid
Infrared spectrum Conforms
Melting point 127 to 134 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR