N-Methylpyrrole-3-boronic acid pinacol ester 90%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-METHYL-1H-PYRROL-3-YLBORONIC ACID PINACOL ESTER, 1H-Pyrrole,1-methyl-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-, 1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole

44100-0010 44100-0050
TS44100-0010EA 260 USD
TS44100-0010 TS44100-0050
N-Methylpyrrole-3-boronic acid pinacol ester 90%
N-Methylpyrrole-3-boronic acid pinacol ester
Formula: C₁₁H₁₈BNO₂
Melting Pt: 44…51 °C
Storage Temperature: Ambient
MDL Number: MFCD13182200
CAS Number: 953040-54-5

Order Now

Specification Test Results

Appearance White to light brown Low melting solid
Infrared spectrum Conforms
GC ≥88.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR