6-Quinolineboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

43964-0010 43964-0050
TS43964-0010EA 79 USD
TS43964-0010 TS43964-0050
6-Quinolineboronic acid pinacol ester 97%
6-Quinolineboronic acid pinacol ester
Formula: C₁₅H₁₈BNO₂
Melting Pt: 65…67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06411327
CAS Number: 406463-06-7

Order Now

Specification Test Results

Appearance White to light orange Powder or crystalline solid
Infrared spectrum Conforms
Melting point 62 to 70 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR