(R)-1-Methyl-3-pyrrolidinol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-3-Hydroxy-1-methylpyrrolidine, (R)-(-)-1-Methyl-3-pyrrolidinol, (R)-3-Hydroxy-1-methyl-pyrrolidine

43944-0010 43944-0050
TS43944-0010EA 114 USD
TS43944-0010 TS43944-0050
(R)-1-Methyl-3-pyrrolidinol 99%
(R)-1-Methyl-3-pyrrolidinol
Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 50…52 °C (1 mmHg)
Density: 0.921 g/cm³
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD03788747
CAS Number: 104641-60-3

Order Now

Specification Test Results

Appearance Colorless to yellow to brown Liquid (May darken on storage)
Infrared spectrum Conforms
Specific optical rotation -6° to -8° (20°C, 589 nm) (c=1 in chloroform)
Titration with HClO4 98.5 to 101.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR