(S)-1-Methyl-3-pyrrolidinol 98%, ee 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-3-Hydroxy-1-methylpyrrolidine, (S)-(+)-1-Methyl-3-pyrrolidinol

43932-0010 43932-0050
TS43932-0010EA 114 USD
TS43932-0010 TS43932-0050
(S)-1-Methyl-3-pyrrolidinol 98%, ee 99%
(S)-1-Methyl-3-pyrrolidinol
Formula: C₅H₁₁NO
Boiling Pt: 181…182 °C (760 mmHg)
Density: 0.993 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD03788748
CAS Number: 104641-59-0

Order Now

Specification Test Results

Appearance Colorless to yellow to brown Liquid (May darken on storage)
Infrared spectrum Conforms
Water ≤2.00 %
GC ≥97.5 %
Specific optical rotation +5° to +7° (20°C, 589 nm) (c=1 in chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR