5-Iodo-2-methoxypyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

43893-0010 43893-0050
TS43893-0010EA 139 USD
TS43893-0010 TS43893-0050
5-Iodo-2-methoxypyridine 97%
5-Iodo-2-methoxypyridine
Formula: C₆H₆INO
Boiling Pt: 106…108 °C (30Torr)
Storage Temperature: Ambient
MDL Number: MFCD07781180
CAS Number: 13472-61-2

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR