2-Amino-2-thiazoline 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,5-Dihydrothiazol-2-amine

43885-0050 43885-1000
TS43885-0050EA 19 USD
TS43885-0050 TS43885-1000
2-Amino-2-thiazoline 97%
2-Amino-2-thiazoline
Formula: C₃H₆N₂S
MW: 102.16 g/mol
Boiling Pt: 221.8 °C (760 mmHg)
Melting Pt: 77…81 °C
Flash Pt: 87.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00005313
CAS Number: 1779-81-3

Order Now

Specification Test Results

Appearance White to off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 78 to 83 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR