3-Ethynylthiophene 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
43858-0010
43858-0050
TS43858-0010EA
79
USD
TS43858-0010
TS43858-0050
3-Ethynylthiophene 96%
3-Ethynylthiophene
Formula:
C₆H₄S Boiling Pt: 152…154 °C (760 mmHg) Density: 1.098 g/cm³ Flash Pt: 46 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04039973 CAS Number: 67237-53-0 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Colorless to yellow to orange Liquid (May darken on storage) |
Infrared spectrum | Conforms |
GC | ≥95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...