N-Boc-pyrrole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
43851-0250
43851-1000
TS43851-0250EA
73
USD
TS43851-0250
TS43851-1000
N-Boc-pyrrole 98%
N-Boc-pyrrole
Formula:
C₉H₁₃NO₂ Boiling Pt: 97 °C (2.3Torr) Density: 1 g/cm³ Flash Pt: 75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00209559 CAS Number: 5176-27-2 |
Specification Test Results
Appearance | Clear yellow to brown Liquid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...