7-Chloropyrazolo[1,5-a]pyrimidine 95%

Supplier: Thermo Scientific Chemicals
Danger

43849-0010 43849-0050
TS43849-0010EA 256 USD
TS43849-0010 TS43849-0050
7-Chloropyrazolo[1,5-a]pyrimidine 95%
7-Chloropyrazolo[1,5-a]pyrimidine
Formula: C₆H₄ClN₃
MW: 153.5700 g/mol
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD04035684
CAS Number: 58347-49-2
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to yellow Solid (May have a pink hue)
Infrared spectrum Conforms
Melting point 99 to 103 °C
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR