1-Boc-4-(2-chloroethyl)piperazine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(2-Chloroethyl)piperazine,N1-BOC protected, tert-Butyl 4-(2-chloroethyl)piperazine-1-carboxylate

43824-0010 43824-0050
TS43824-0010EA 128 USD
TS43824-0010 TS43824-0050
1-Boc-4-(2-chloroethyl)piperazine 97%
1-Boc-4-(2-chloroethyl)piperazine
Formula: C₁₁H₂₁ClN₂O₂
MW: 248.751 g/mol
Melting Pt: 62.9…64.7 °C
Storage Temperature: Ambient
MDL Number: MFCD08443993
CAS Number: 208167-83-3

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 62 to 68 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR