Methyl picolinate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl pyridine-2-carboxylate

43814-0050 43814-0250 43814-1000
TS43814-0050EA 41 USD
TS43814-0050 TS43814-0250 TS43814-1000
Methyl picolinate 98%
Methyl picolinate
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 95 °C (1hPa)
Melting Pt: 14…16 °C
Density: 1.137 g/cm³
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00038038
CAS Number: 2459-07-6

Order Now

Specification Test Results

Appearance Clear colorless to yellow to orange Liquid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR