4-Bromo-3-(trifluoromethyl)-1H-pyrazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromo-3-(trifluoromethyl)-1H-pyrazole

43804-0010 43804-0050
TS43804-0010EA 181 USD
TS43804-0010 TS43804-0050
4-Bromo-3-(trifluoromethyl)-1H-pyrazole 97%
4-Bromo-3-(trifluoromethyl)-1H-pyrazole
Formula: C₄H₂BrF₃N₂
MW: 214.97 g/mol
Melting Pt: 112…114 °C
Storage Temperature: Ambient
MDL Number: MFCD04969230
CAS Number: 19968-17-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 111 to 115 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR