5-Amino-2,3-dihydrobenzo[b]furan 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,3-Dihydrobenzofuran-5-amine, 5-Amino-2,3-dihydrobenzofuran, 2,3-dihydro-1-benzofuran-5-amine, 5-Benzofuranamine,2,3-dihydro-

43803-0010 43803-0050
TS43803-0010EA 185 USD
TS43803-0010 TS43803-0050
5-Amino-2,3-dihydrobenzo[b]furan 97%
5-Amino-2,3-dihydrobenzo[b]furan
Formula: C₈H₉NO
MW: 135.166 g/mol
Melting Pt: 81.6…83.4 °C
Storage Temperature: Ambient
MDL Number: MFCD03617969
CAS Number: 42933-43-7

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
Melting point 80 to 84 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR