tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
43798-0010
43798-0050
TS43798-0010EA
186
USD
TS43798-0010
TS43798-0050
tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate 97%
tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate
Formula:
C₁₁H₁₉NO₃ Melting Pt: 58…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD07779385 CAS Number: 147804-30-6 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 55 to 60 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...