2-Methyl-1,3-benzoxazol-5-amine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Amino-2-methyl-1,3-benzoxazole, 2-Methylbenzo[d]oxazol-5-amine

43792-0010 43792-0050
TS43792-0010EA 137 USD
TS43792-0010 TS43792-0050
2-Methyl-1,3-benzoxazol-5-amine 97%
2-Methyl-1,3-benzoxazol-5-amine
Formula: C₈H₈N₂O
MW: 148.161 g/mol
Melting Pt: 72…74.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00628970
CAS Number: 72745-76-7

Order Now

Specification Test Results

Appearance Off-white to brown Crystalline powder
Infrared spectrum Conforms
Melting point 70 to 76 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR