2-Amino-3-bromo-5-chloropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

43791-0010 43791-0050
TS43791-0010EA 58 USD
TS43791-0010 TS43791-0050
2-Amino-3-bromo-5-chloropyridine 97%
2-Amino-3-bromo-5-chloropyridine
Formula: C₅H₄BrClN₂
Melting Pt: 82…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00955627
CAS Number: 26163-03-1

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 83 to 87 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR