(S)-(+)-3-Amino-1-boc-piperidine 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
43749-0010
43749-0050
TS43749-0010EA
252
USD
TS43749-0010
TS43749-0050
(S)-(+)-3-Amino-1-boc-piperidine 95%
(S)-(+)-3-Amino-1-Boc-piperidine
Formula:
C₁₀H₂₀N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD03094718 CAS Number: 625471-18-3 UN: 2735 ADR: 8,III |
Specification Test Results
Appearance | Colorless to yellow Gum or semi-solid |
Infrared spectrum | Conforms |
GC | ≥94.0 % |
Specific optical rotation | +26.5° to +30.5° (20°C, 589 nm) (c=1 in DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...