(S)-3-(Aminomethyl)-1-boc-piperidine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
43747-0010
43747-0050
TS43747-0010EA
192
USD
TS43747-0010
TS43747-0050
(S)-3-(Aminomethyl)-1-boc-piperidine 97%
(S)-3-(Aminomethyl)-1-Boc-piperidine
Formula:
C₁₁H₂₂N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD03839877 CAS Number: 140645-24-5 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Specific optical rotation | -10° to -20° (20°C, 589 nm) (c=1, methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...