4-Iodonicotinonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Cyano-4-iodopyridine

43745-0010 43745-0050
TS43745-0010EA 430 USD
TS43745-0010 TS43745-0050
4-Iodonicotinonitrile 97%
4-Iodonicotinonitrile
Formula: C₆H₃IN₂
MW: 230.01
Melting Pt: 114…115 °C
Storage Temperature: Freezer
MDL Number: MFCD09037453
CAS Number: 490039-72-0

Order Now

Specification Test Results

Appearance Light brown Crystalline powder
Infrared spectrum Conforms
Melting point 112 to 116 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR