4-Amino-2-bromopyridine 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Bromo-4-pyridylamine, 2-Bromopyridin-4-amine

43741-0010 43741-0050
TS43741-0010EA 32 USD
TS43741-0010 TS43741-0050
4-Amino-2-bromopyridine 95%
4-Amino-2-bromopyridine
Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD01646061
CAS Number: 7598-35-8

Order Now

Specification Test Results

Appearance White to orange to light brown Powder
Infrared spectrum Conforms
HPLC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR